accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18156-74-6

IUPAC Name: 1-(trimethylsilyl)-1H-imidazole
Synonyms: 1-(trimethylsilyl)imidazole
Molecular Weight (g/mol): 140.26
Molecular Formula: C6H12N2Si
InChi Key: YKFRUJSEPGHZFJ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N1C=CN=C1
Boiling Point 
  • (4)
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)