accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18166-64-8

IUPAC Name: (2E)-3-(4-chlorophenyl)prop-2-enamide
Synonyms: (2E)-3-(4-chlorophenyl)prop-2-enamide
Molecular Weight (g/mol): 181.62
Molecular Formula: C9H8ClNO
InChi Key: PWXPFYVNYKVJBW-ZZXKWVIFSA-N
SMILES: NC(=O)\C=C\C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results