accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNOO

CAS RN 18190-44-8

IUPAC Name: 1-(2-hydroxyethyl)pyrrolidine-2,5-dione
Synonyms: 1-(2-hydroxyethyl)pyrrolidine-2,5-dione
Molecular Weight (g/mol): 143.14
Molecular Formula: C6H9NO3
InChi Key: TWYIPMITVXPNEM-UHFFFAOYSA-N
SMILES: OCCN1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results