CAS RN 182004-65-5
CAS RN 182004-65-5
IUPAC Name:
[(2-{4-[(4-nitrophenyl)methoxy]phenyl}ethyl)sulfanyl]methanimidamide; methanesulfonic acid
Synonyms:
(2-{4-[(4-nitrophenyl)methoxy]phenyl}ethyl)sulfanylmethanimidamide; methanesulfonic acid
Molecular Weight (g/mol):
427.49
Molecular Formula:
C17H21N3O6S2
InChi Key:
WGIKEBHIKKWJLG-UHFFFAOYSA-N
SMILES:
CS(O)(=O)=O.NC(=N)SCCC1=CC=C(OCC2=CC=C(C=C2)[N+]([O-])=O)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ KB-R7943 mesylate
CAS: 182004-65-5 Molecular Formula: C17H21N3O6S2