accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18202-73-8

IUPAC Name: hydrogen 2,2-dimethylpropanimidamide chloride
Synonyms: hydrogen 2,2-dimethylpropanimidamide chloride
Molecular Weight (g/mol): 136.62
Molecular Formula: C5H13ClN2
InChi Key: ARDGQYVTLGUJII-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(C)C(N)=N
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)