accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNSOO

CAS RN 18212-21-0

IUPAC Name: 4-methyl-1,2,3-thiadiazole-5-carboxylate
Synonyms: 4-methyl-1,2,3-thiadiazole-5-carboxylate
Molecular Weight (g/mol): 143.14
Molecular Formula: C4H3N2O2S
InChi Key: NHHQOYLPBUYHQU-UHFFFAOYSA-M
SMILES: CC1=C(SN=N1)C([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)