accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SNNCl

CAS RN 182198-35-2

IUPAC Name: 4-chloro-5-phenylthieno[2,3-d]pyrimidine
Synonyms: 4-chloro-5-phenylthieno[2,3-d]pyrimidine
Molecular Weight (g/mol): 246.71
Molecular Formula: C12H7ClN2S
InChi Key: WONOKVSIDWOIGC-UHFFFAOYSA-N
SMILES: ClC1=C2C(SC=C2C2=CC=CC=C2)=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results