accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1823-14-9

HC

CAS RN 1823-14-9

IUPAC Name: (pent-4-yn-1-yl)benzene
Synonyms: pent-4-yn-1-ylbenzene
Molecular Weight (g/mol): 144.22
Molecular Formula: C11H12
InChi Key: KOSORCNALVBYBP-UHFFFAOYSA-N
SMILES: C#CCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results