CAS RN 1824308-55-5
IUPAC Name:
(5-bromo-3-methylpyridin-2-yl)thiourea
Synonyms:
5-bromo-3-methylpyridin-2-ylthiourea
Molecular Weight (g/mol):
246.13
Molecular Formula:
C7H8BrN3S
InChi Key:
OGQFHROQBUNGAO-UHFFFAOYSA-N
SMILES:
CC1=CC(Br)=CN=C1NC(N)=S
1
–
1
of
1
results
1
N-(5-Bromo-3-methyl-2-pyridyl)thiourea, 97%, Thermo Scientific™
CAS: 1824308-55-5 Molecular Formula: C7H8BrN3S Molecular Weight (g/mol): 246.126 MDL Number: MFCD20921516 InChI Key: OGQFHROQBUNGAO-UHFFFAOYSA-N Synonym: n-5-bromo-3-methyl-2-pyridyl thiourea PubChem CID: 96562247 IUPAC Name: (5-bromo-3-methylpyridin-2-yl)thiourea SMILES: CC1=CC(=CN=C1NC(=S)N)Br