accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH2MgBr

CAS RN 1826-67-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 131.26
Molecular Formula: C2H3BrMg
InChi Key: RMGJCSHZTFKPNO-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].C=[CH-]
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results