accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBrOHO

CAS RN 18263-25-7

IUPAC Name: 2-bromohexadecanoic acid
Synonyms: 2-bromohexadecanoic acid
Molecular Weight (g/mol): 335.33
Molecular Formula: C16H31BrO2
InChi Key: DPRAYRYQQAXQPE-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCC(Br)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results