CAS RN 18282-10-5
Tin(IV) oxide, Puratronic™, 99.996% (metals basis)
CAS: 18282-10-5 Molecular Formula: O2Sn Molecular Weight (g/mol): 150.71 MDL Number: MFCD00011244 InChI Key: XOLBLPGZBRYERU-UHFFFAOYSA-N Synonym: tin dioxide,tin iv oxide,stannic oxide,cassiterite,flowers of tin,stannic dioxide,tin oxide sno2,tin peroxide,white tin oxide,stannic anhydride PubChem CID: 29011 ChEBI: CHEBI:52991 IUPAC Name: dioxotin SMILES: O=[Sn]=O
Tin(IV) oxide, 99.9%, (trace metal basis)
CAS: 18282-10-5 Molecular Formula: O2Sn Molecular Weight (g/mol): 150.71 MDL Number: MFCD00011244 InChI Key: XOLBLPGZBRYERU-UHFFFAOYSA-N Synonym: tin dioxide,tin iv oxide,stannic oxide,cassiterite,flowers of tin,stannic dioxide,tin oxide sno2,tin peroxide,white tin oxide,stannic anhydride PubChem CID: 29011 ChEBI: CHEBI:52991 IUPAC Name: stannanedione SMILES: O=[Sn]=O
Tin(IV) oxide, 99.9% (metals basis)
CAS: 18282-10-5 Molecular Formula: O2Sn Molecular Weight (g/mol): 150.71 MDL Number: MFCD00011244 InChI Key: XOLBLPGZBRYERU-UHFFFAOYSA-N Synonym: tin dioxide,tin iv oxide,stannic oxide,cassiterite,flowers of tin,stannic dioxide,tin oxide sno2,tin peroxide,white tin oxide,stannic anhydride PubChem CID: 29011 ChEBI: CHEBI:52991 SMILES: O=[Sn]=O
Tin(IV) oxide, 99%, pure
CAS: 18282-10-5 Molecular Formula: O2Sn Molecular Weight (g/mol): 150.71 MDL Number: MFCD00011244 InChI Key: XOLBLPGZBRYERU-UHFFFAOYSA-N Synonym: tin dioxide,tin iv oxide,stannic oxide,cassiterite,flowers of tin,stannic dioxide,tin oxide sno2,tin peroxide,white tin oxide,stannic anhydride PubChem CID: 29011 ChEBI: CHEBI:52991 IUPAC Name: dioxotin SMILES: O=[Sn]=O
Tin(IV) oxide, 99.9% (metals basis)
CAS: 18282-10-5 Molecular Formula: O2Sn Molecular Weight (g/mol): 150.71 MDL Number: MFCD00011244 InChI Key: XOLBLPGZBRYERU-UHFFFAOYSA-N Synonym: tin dioxide,tin iv oxide,stannic oxide,cassiterite,flowers of tin,stannic dioxide,tin oxide sno2,tin peroxide,white tin oxide,stannic anhydride PubChem CID: 29011 ChEBI: CHEBI:52991 IUPAC Name: dioxotin SMILES: O=[Sn]=O
Tin oxide, polymeric precursor, Oxide ≈29 wt%, Thermo Scientific™
CAS: 18282-10-5 Molecular Formula: O2Sn Molecular Weight (g/mol): 150.71 MDL Number: MFCD00011244 InChI Key: XOLBLPGZBRYERU-UHFFFAOYSA-N Synonym: tin dioxide,tin iv oxide,stannic oxide,cassiterite,flowers of tin,stannic dioxide,tin oxide sno2,tin peroxide,white tin oxide,stannic anhydride PubChem CID: 29011 ChEBI: CHEBI:52991 IUPAC Name: dioxotin SMILES: O=[Sn]=O
Tin(IV) oxide, 15% in H{2}O colloidal dispersion
CAS: 18282-10-5 Molecular Formula: O2Sn Molecular Weight (g/mol): 150.71 MDL Number: MFCD00011244 InChI Key: XOLBLPGZBRYERU-UHFFFAOYSA-N Synonym: tin dioxide,tin iv oxide,stannic oxide,cassiterite,flowers of tin,stannic dioxide,tin oxide sno2,tin peroxide,white tin oxide,stannic anhydride PubChem CID: 29011 ChEBI: CHEBI:52991 IUPAC Name: stannanedione SMILES: O=[Sn]=O