CAS RN 18312-57-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-Bis(2-hydroxyethyl) Tridecylamine, TRC
CAS: 18312-57-7 Molecular Formula: C17H37NO2 Molecular Weight (g/mol): 287.48 IUPAC Name: 2-[2-hydroxyethyl(tridecyl)amino]ethanol SMILES: CCCCCCCCCCCCCN(CCO)CCO