accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 183133-96-2

IUPAC Name: 4-(acetyloxy)-15-[(3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-3-phenylpropanoyl)oxy]-1-hydroxy-9,12-dimethoxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
Synonyms: cabazitaxel
Molecular Weight (g/mol): 835.94
Molecular Formula: C45H57NO14
InChi Key: BMQGVNUXMIRLCK-UHFFFAOYNA-N
SMILES: COC1CC2OCC2(OC(C)=O)C2C(OC(=O)C3=CC=CC=C3)C3(O)CC(OC(=O)C(O)C(NC(=O)OC(C)(C)C)C4=CC=CC=C4)C(C)=C(C(OC)C(=O)C12C)C3(C)C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results