accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BOHHOH3C

CAS RN 183158-34-1

IUPAC Name: (2,3-dimethylphenyl)boronic acid
Synonyms: 2,3-dimethylphenylboronic acid
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: ZYYANAWVBDFAHY-UHFFFAOYSA-N
SMILES: CC1=CC=CC(B(O)O)=C1C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)