accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHOONOO

CAS RN 18342-66-0

IUPAC Name: 2,5-dioxopyrrolidin-1-yl N-methylcarbamate
Synonyms: 2,5-dioxopyrrolidin-1-yl N-methylcarbamate
Molecular Weight (g/mol): 172.14
Molecular Formula: C6H8N2O4
InChi Key: XMNGSPOWUCNRMO-UHFFFAOYSA-N
SMILES: CNC(=O)ON1C(=O)CCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results