accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 183438-24-6

IUPAC Name: 5-bromo-2-iodopyrimidine
Synonyms: 5-bromo-2-iodopyrimidine
Molecular Weight (g/mol): 284.88
Molecular Formula: C4H2BrIN2
InChi Key: ZEZKXPQIDURFKA-UHFFFAOYSA-N
SMILES: BrC1=CN=C(I)N=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results