accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOClCl

CAS RN 1836-75-5

IUPAC Name: 2,4-dichloro-1-(4-nitrophenoxy)benzene
Synonyms: nitrofen
Molecular Weight (g/mol): 284.09
Molecular Formula: C12H7Cl2NO3
InChi Key: XITQUSLLOSKDTB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC2=CC=C(Cl)C=C2Cl)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Solvent or Matrix 
  • (2)
  • (2)
  • (1)
Concentration 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results