CAS RN 183677-71-6
IUPAC Name:
2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Synonyms:
2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol):
268.95
Molecular Formula:
C11H14BBrO2
InChi Key:
GUASPWAWPYERCR-UHFFFAOYSA-N
SMILES:
CC1(C)COB(OC1)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
4-Bromobenzeneboronic acid neopentyl glycol ester, 98+%
CAS: 183677-71-6 Molecular Formula: C11H14BBrO2 Molecular Weight (g/mol): 268.945 MDL Number: MFCD01318105 InChI Key: GUASPWAWPYERCR-UHFFFAOYSA-N Synonym: 2-4-bromophenyl-5,5-dimethyl-1,3,2-dioxaborinane,4-bromobenzeneboronic acid neopentyl glycol ester,4-bromophenylboronic acid neopentyl glycol ester,amtb980,2-4-bromophenyl-5,5-dimethyl-2-bora-1,3-dioxane,4-bromophenylboronic acid neopentyl glycol cyclic ester PubChem CID: 10400865 IUPAC Name: 2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane SMILES: B1(OCC(CO1)(C)C)C2=CC=C(C=C2)Br