accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ClN

CAS RN 18368-64-4

IUPAC Name: 2-chloro-5-methylpyridine
Synonyms: 2-chloro-5-methylpyridine
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: VXLYOURCUVQYLN-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)N=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)