accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOHN

CAS RN 18372-22-0

IUPAC Name: methyl 2-(1H-indol-3-yl)-2-oxoacetate
Synonyms: methyl 2-(1H-indol-3-yl)-2-oxoacetate
Molecular Weight (g/mol): 203.2
Molecular Formula: C11H9NO3
InChi Key: VFIJGAWYVXDYLK-UHFFFAOYSA-N
SMILES: COC(=O)C(=O)C1=CNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results