accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrHOCl

CAS RN 183802-98-4

IUPAC Name: 5-bromo-2-chlorophenol
Synonyms: 5-bromo-2-chlorophenol
Molecular Weight (g/mol): 207.45
Molecular Formula: C6H4BrClO
InChi Key: UEVFFMZHGNYDKM-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results