accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOFFFFFFClO

CAS RN 18381-53-8

IUPAC Name: ethyl 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxopentanoate
Synonyms: ethyl 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxopentanoate
Molecular Weight (g/mol): 286.55
Molecular Formula: C7H5ClF6O3
InChi Key: OLRXGDHRDQKNGW-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results