accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18486-69-6

H3COH3C

CAS RN 18486-69-6

IUPAC Name: 6,6-dimethylbicyclo[3.1.1]hept-2-ene-2-carbaldehyde
Synonyms: myrtenal
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: KMRMUZKLFIEVAO-UHFFFAOYNA-N
SMILES: CC1(C)C2CC1C(C=O)=CC2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results