accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHHOOHH2O

CAS RN 184901-84-6

IUPAC Name: 2-hydroxy-2-(4-hydroxyphenyl)acetic acid hydrate
Synonyms: 4-hydroxymandelic acid hydrate
Molecular Weight (g/mol): 186.16
Molecular Formula: C8H10O5
InChi Key: ATPBHLAWGXOMOR-UHFFFAOYNA-N
SMILES: O.OC(C(O)=O)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results