accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18497-13-7

H2OH2OH2OH2OH2OH2OClPtClClClClClH3OH3O

CAS RN 18497-13-7

IUPAC Name: dihydrogen hexachloroplatinumdiuide hexahydrate
Synonyms: dihydrogen hexachloroplatinate(2-) hexahydrate
Molecular Weight (g/mol): 523.94
Molecular Formula: Cl6H20O6Pt
InChi Key: RHSKGSPRYBMZLA-UHFFFAOYSA-P
SMILES: [H+].[H+].O.O.O.O.O.O.Cl.Cl.Cl.Cl.Cl.Cl.[Pt--]
Molecular Weight (g/mol) 
  • (13)
  • (3)
Quantity 
  • (6)
  • (3)
  • (1)
  • (6)
Grade 
  • (4)
  • (4)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (6)
  • (3)
  • (13)
  • (3)
  • (6)
  • (3)
  • (1)
  • (6)
  • (4)
  • (4)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)