accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 184970-25-0

FFFFBr

CAS RN 184970-25-0

IUPAC Name: 1-(bromomethyl)-2-fluoro-3-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-2-fluoro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: QBEHXDXQUVMEQX-UHFFFAOYSA-N
SMILES: FC1=C(CBr)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results