accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3ON

CAS RN 185259-36-3

IUPAC Name: 4-(tert-butoxy)benzonitrile
Synonyms: 4-(tert-butoxy)benzonitrile
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: WLWYCCYGVDZFMS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=CC=C(C=C1)C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results