accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18532-87-1

NNNClNClClRuN

CAS RN 18532-87-1

IUPAC Name: ruthenium(3+) pentaamine trichloride
Synonyms: ruthenium(3+) pentaamine trichloride
Molecular Weight (g/mol): 292.58
Molecular Formula: Cl3H15N5Ru
InChi Key: HLAVJMYZYQTXAH-UHFFFAOYSA-K
SMILES: N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results