accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3NHSOOOCH3

CAS RN 1858242-03-1

IUPAC Name: N-[(3-tert-butylphenyl)methyl]-4-methoxybenzene-1-sulfonamide
Synonyms: N-[(3-tert-butylphenyl)methyl]-4-methoxybenzenesulfonamide
Molecular Weight (g/mol): 333.45
Molecular Formula: C18H23NO3S
InChi Key: CHWBOLSVGNXPNI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)S(=O)(=O)NCC1=CC(=CC=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results