accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1858255-71-6

NOCl

CAS RN 1858255-71-6

IUPAC Name: N-benzyl-4-(chloromethyl)-N-phenylbenzamide
Synonyms: N-benzyl-4-(chloromethyl)-N-phenylbenzamide
Molecular Weight (g/mol): 335.83
Molecular Formula: C21H18ClNO
InChi Key: ITWJOMOXAADOGB-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(C=C1)C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results