accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18595-12-5

IUPAC Name: methyl 5-amino-2-methylbenzoate
Synonyms: methyl 5-amino-2-methylbenzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: JNPZKGOLYSCSEL-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results