accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18595-18-1

IUPAC Name: methyl 3-amino-4-methylbenzoate
Synonyms: methyl 3-amino-4-methylbenzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: YEPWCJHMSVABPQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)C(N)=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results