accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18598-63-5

IUPAC Name: hydrogen methyl (2R)-2-amino-3-sulfanylpropanoate chloride
Synonyms: hydrogen cysteine methyl ester chloride
Molecular Weight (g/mol): 171.64
Molecular Formula: C4H10ClNO2S
InChi Key: WHOHXJZQBJXAKL-DFWYDOINSA-N
SMILES: [H+].[Cl-].COC(=O)[C@@H](N)CS
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)