accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO(S)SNHOO

CAS RN 186320-06-9

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(thiophen-3-yl)propanoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(thiophen-3-yl)propanoate
Molecular Weight (g/mol): 392.45
Molecular Formula: C22H18NO4S
InChi Key: LSBZJMRHROCYGY-FQEVSTJZSA-M
SMILES: [O-]C(=O)[C@H](CC1=CSC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)