accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNONN

CAS RN 18637-83-7

IUPAC Name: bis(1H-imidazol-1-yl)ethane-1,2-dione
Synonyms: bis(imidazol-1-yl)ethane-1,2-dione
Molecular Weight (g/mol): 190.16
Molecular Formula: C8H6N4O2
InChi Key: ONRNRVLJHFFBJG-UHFFFAOYSA-N
SMILES: O=C(N1C=CN=C1)C(=O)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results