accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ICH3OSNCH3CH3H3C

CAS RN 1866-15-5

IUPAC Name: [2-(acetylsulfanyl)ethyl]trimethylazanium iodide
Synonyms: [2-(acetylsulfanyl)ethyl]trimethylazanium iodide
Molecular Weight (g/mol): 289.18
Molecular Formula: C7H16INOS
InChi Key: NTBLZMAMTZXLBP-UHFFFAOYSA-M
SMILES: [I-].CC(=O)SCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (5)
  • (3)