accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18672-03-2

ClClNHNH2N

CAS RN 18672-03-2

IUPAC Name: 5,6-dichloro-1H-1,3-benzodiazol-2-amine
Synonyms: 5,6-dichloro-1H-1,3-benzodiazol-2-amine
Molecular Weight (g/mol): 202.04
Molecular Formula: C7H5Cl2N3
InChi Key: HAADRTMPUSJNOI-UHFFFAOYSA-N
SMILES: NC1=NC2=CC(Cl)=C(Cl)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results