accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOHNHN(Z)

CAS RN 18704-67-1

IUPAC Name: 4-chloro-6-[(3Z)-2,3-dihydro-1H-pyrazol-3-ylidene]cyclohexa-2,4-dien-1-one
Synonyms: 4-chloro-6-[(3Z)-1,2-dihydropyrazol-3-ylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 194.62
Molecular Formula: C9H7ClN2O
InChi Key: BAWPNSUPRVQKKN-FPLPWBNLSA-N
SMILES: ClC1=C\C(=C2\NNC=C2)C(=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results