accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18717-73-2

IUPAC Name: 8-benzyl-8-azabicyclo[3.2.1]octan-3-ol
Synonyms: 8-benzyl-8-azabicyclo[3.2.1]octan-3-ol
Molecular Weight (g/mol): 217.31
Molecular Formula: C14H19NO
InChi Key: HCBGIBWAPOFRKI-UHFFFAOYNA-N
SMILES: OC1CC2CCC(C1)N2CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results