accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 187237-37-2

IUPAC Name: tert-butyl N-[5-(aminomethyl)pyridin-2-yl]carbamate
Synonyms: tert-butyl N-[5-(aminomethyl)pyridin-2-yl]carbamate
Molecular Weight (g/mol): 223.28
Molecular Formula: C11H17N3O2
InChi Key: DFLQTVPEIMTXSZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results