accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSiH3CCH3SiHSiCH3CH3H3CSiCH3CH3H3C

CAS RN 1873-77-4

IUPAC Name: 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
Synonyms: 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
Molecular Weight (g/mol): 248.66
Molecular Formula: C9H28Si4
InChi Key: SQMFULTZZQBFBM-UHFFFAOYSA-N
SMILES: C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)