accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18735-74-5

IUPAC Name: 5-methyl-2-phenyl-1,3-oxazole-4-carboxylate
Synonyms: 5-methyl-2-phenyl-1,3-oxazole-4-carboxylate
Molecular Weight (g/mol): 202.19
Molecular Formula: C11H8NO3
InChi Key: YABCPNYCFFUVNM-UHFFFAOYSA-M
SMILES: CC1=C(N=C(O1)C1=CC=CC=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results