accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1877-75-4

IUPAC Name: 2-(4-methoxyphenoxy)acetic acid
Synonyms: P-methoxyphenoxyacetic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: BHFSBJHPPFJCOS-UHFFFAOYSA-N
SMILES: COC1=CC=C(OCC(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results