accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1877-77-6

IUPAC Name: (3-aminophenyl)methanol
Synonyms: (3-aminophenyl)methanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: OJZQOQNSUZLSMV-UHFFFAOYSA-N
SMILES: NC1=CC=CC(CO)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results