accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1878-68-8

IUPAC Name: 2-(4-bromophenyl)acetic acid
Synonyms: C8H7BrO2 P-bromophenylacetic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: QOWSWEBLNVACCL-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)