accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1878-71-3

OOHN

CAS RN 1878-71-3

IUPAC Name: 2-(3-cyanophenyl)acetic acid
Synonyms: M-cyanophenylacetic acid
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: ZNXMNKGBHYVNNE-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(=C1)C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results