accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOONOO

CAS RN 1878-87-1

IUPAC Name: 2-(2-nitrophenoxy)acetic acid
Synonyms: O-nitrophenoxyacetic acid
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: TYHHDWAHJRRYCU-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results