accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SH3C

CAS RN 18800-53-8

IUPAC Name: (3,4-dimethylphenyl)sulfanide
Synonyms: (3,4-dimethylphenyl)sulfanide
Molecular Weight (g/mol): 137.22
Molecular Formula: C8H9S
InChi Key: IDKCKPBAFOIONK-UHFFFAOYSA-M
SMILES: CC1=CC=C([S-])C=C1C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)