accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NHNNN(R)(S)HONNH2NHNNN(R)(S)HONOHSOHOO

CAS RN 188062-50-2

IUPAC Name: bis([(1S,4R)-4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopent-2-en-1-yl]methanol); sulfuric acid
Synonyms: abacavir sulfate
Molecular Weight (g/mol): 670.75
Molecular Formula: C28H38N12O6S
InChi Key: WMHSRBZIJNQHKT-FFKFEZPRSA-N
SMILES: OS(O)(=O)=O.NC1=NC(NC2CC2)=C2N=CN([C@@H]3C[C@H](CO)C=C3)C2=N1.NC1=NC(NC2CC2)=C2N=CN([C@@H]3C[C@H](CO)C=C3)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results